The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282923.1 | NP_001269852.1 | 5 - 6 | 560 | 73 |
NM_017790.4 | NP_060260.3 | 5 - 6 | 454 | 73 | |
NM_144488.5 | NP_652759.3 | 8 - 9 | 898 | 73 | |
XM_006717219.2 | XP_006717282.1 | 7 - 8 | 744 | 73 | |
XM_006717220.1 | XP_006717283.1 | 5 - 6 | 520 | 73 | |
XM_011518893.1 | XP_011517195.1 | 8 - 9 | 896 | 73 | |
XM_017015003.1 | XP_016870492.1 | 8 - 9 | 896 | 73 | |
GenBank mRNA | AF490839.1 | - | 5 - 6 | 375 | 73 |
AK074977.1 | - | 5 - 6 | 424 | 73 | |
AK125059.1 | - | 5 - 6 | 531 | 73 | |
AK128127.1 | - | 8 - 9 | 896 | 73 | |
DA680346.1 | - | 5 - 6 | 454 | 73 |