The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278730.1 | NP_001265659.1 | 4 - 5 | 1019 | 97 |
NM_006478.4 | NP_006469.2 | 4 - 5 | 1047 | 97 | |
NM_152236.2 | NP_689422.1 | 4 - 5 | 1052 | 97 | |
XM_011529824.1 | XP_011528126.1 | 5 - 6 | 1171 | 97 | |
XM_011529825.1 | XP_011528127.1 | 4 - 5 | 980 | 97 | |
XM_017028533.1 | XP_016884022.1 | 4 - 5 | 1697 | 97 | |
GenBank mRNA | AK093386.1 | - | 4 - 5 | 989 | 97 |
AK122829.1 | - | 3 - 4 | 1889 | 97 | |
AK223602.1 | - | 4 - 5 | 991 | 97 | |
AK225484.1 | - | 4 - 5 | 991 | 97 | |
AK301715.1 | - | 3 - 4 | 474 | 97 | |
BC001782.1 | - | 4 - 5 | 987 | 97 | |
BC007624.1 | - | 4 - 5 | 987 | 97 | |
BC011047.1 | - | 4 - 5 | 990 | 97 | |
Y07846.1 | - | 4 - 5 | 973 | 97 |