Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CEP70
Entrez Gene ID
80321
Gene Name
centrosomal protein 70
Gene Aliases
BITE
Chromosome Location
Chr.3: 138494339 - 138594383 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320599.1 NP_001307528.1 17 - 18 2026 88
XM_005247805.3 XP_005247862.1 15 - 16 1746 88

View Full Product Details