Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CEP70
Entrez Gene ID
80321
Gene Name
centrosomal protein 70
Gene Aliases
BITE
Chromosome Location
Chr.3: 138494339 - 138594383 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001288964.1 NP_001275893.1 9 - 10 1052 113
NM_001288965.1 NP_001275894.1 8 - 9 996 113
NM_001288966.1 NP_001275895.1 9 - 10 1070 113
NM_001288967.1 NP_001275896.1 5 - 6 600 113
NM_001320598.1 NP_001307527.1 9 - 10 1122 113
NM_001320599.1 NP_001307528.1 9 - 10 1070 113
NM_024491.3 NP_077817.2 9 - 10 1070 113
XM_005247805.3 XP_005247862.1 7 - 8 790 113
XM_011513209.2 XP_011511511.1 7 - 8 790 113
XM_017007270.1 XP_016862759.1 7 - 8 790 113
XM_017007271.1 XP_016862760.1 9 - 10 972 113
XM_017007272.1 XP_016862761.1 9 - 10 1310 113
XM_017007273.1 XP_016862762.1 8 - 9 915 113
XM_017007274.1 XP_016862763.1 8 - 9 1335 113
XM_017007275.1 XP_016862764.1 9 - 10 1406 113
XM_017007276.1 XP_016862765.1 5 - 6 603 113
XM_017007277.1 XP_016862766.1 5 - 6 535 113
XM_017007278.1 XP_016862767.1 10 - 11 1141 113
XM_017007279.1 XP_016862768.1 8 - 9 904 113
XM_017007280.1 XP_016862769.1 7 - 8 745 113
XM_017007281.1 XP_016862770.1 2 - 3 918 113
XM_017007282.1 XP_016862771.1 8 - 9 871 113
XM_017007283.1 XP_016862772.1 8 - 9 802 113
XM_017007284.1 XP_016862773.1 2 - 3 1106 113
GenBank mRNA AF202146.1 - 9 - 10 924 113
AF289495.1 - 9 - 10 924 113
AK023098.1 - 7 - 8 785 113
AK225440.1 - 9 - 10 935 113
AK294587.1 - 5 - 6 475 113
AK299003.1 - 8 - 9 861 113
AK301913.1 - 9 - 10 929 113
AK314314.1 - 9 - 10 1042 113
BC030598.2 - 9 - 10 972 113

View Full Product Details