The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127181.2 | NP_001120653.1 | 1 - 2 | 501 | 70 |
NM_033319.3 | NP_201576.1 | 1 - 2 | 501 | 70 | |
GenBank mRNA | BC007071.1 | - | 1 - 2 | 36 | 70 |
BC033154.1 | - | 1 - 2 | 184 | 70 | |
BC066658.1 | - | 1 - 2 | 183 | 70 | |
DN995099.1 | - | 1 - 2 | 509 | 70 |