Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RNH1
Entrez Gene ID
6050
Gene Name
ribonuclease/angiogenin inhibitor 1
Gene Aliases
RAI, RNH
Chromosome Location
Chr.11: 494512 - 507283 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002939.3 NP_002930.2 10 - 11 1765 158
NM_203383.1 NP_976317.1 10 - 11 1697 158
NM_203384.1 NP_976318.1 9 - 10 1524 158
NM_203385.1 NP_976319.1 9 - 10 1592 158
NM_203386.2 NP_976320.1 10 - 11 1655 158
NM_203387.2 NP_976321.1 10 - 11 1723 158
NM_203388.2 NP_976322.1 9 - 10 1482 158
NM_203389.2 NP_976323.1 9 - 10 1550 158
XM_011520255.1 XP_011518557.1 10 - 11 1700 158
XM_011520257.2 XP_011518559.1 10 - 11 1948 158
XM_011520258.2 XP_011518560.1 10 - 11 1916 158
XM_011520259.2 XP_011518561.1 9 - 10 1465 158
XM_011520260.2 XP_011518562.1 10 - 11 1624 158
XM_011520261.2 XP_011518563.1 10 - 11 1698 158
XM_011520262.2 XP_011518564.1 10 - 11 1881 158
XM_011520263.1 XP_011518565.1 9 - 10 1432 158
XM_017018106.1 XP_016873595.1 10 - 11 1765 158
GenBank mRNA AB209196.1 - 8 - 9 3015 158
AK057144.1 - 1 - 2 1814 158
AK311379.1 - 6 - 7 814 158
AL161967.1 - 8 - 9 2700 158
BC000677.2 - 9 - 10 1452 158
BC003506.1 - 9 - 10 1465 158
BC011186.1 - 8 - 9 1250 158
BC011500.1 - 9 - 10 1403 158
BC014629.2 - 9 - 10 1452 158
BC024037.1 - 9 - 10 1465 158
BC047730.1 - 10 - 11 1744 158
BQ650217.1 - 2 - 3 274 158
M22414.1 - 9 - 10 1413 158
M36717.1 - 10 - 11 1652 158
X13973.1 - 10 - 11 1652 158

View Full Product Details