Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLEKHA3
Entrez Gene ID
65977
Gene Name
pleckstrin homology domain containing A3
Gene Aliases
FAPP1
Chromosome Location
Chr.2: 178480410 - 178516324 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019091.3 NP_061964.3 5 - 6 1015 72
XM_011511689.2 XP_011509991.1 6 - 7 1124 72
GenBank mRNA AF286162.1 - 5 - 6 854 72
AK000074.1 - 3 - 4 418 72
AK074209.1 - 5 - 6 990 72
AK314459.1 - 5 - 6 1019 72
BC044567.1 - 5 - 6 709 72
DQ786318.1 - 1 - 2 172 72

View Full Product Details