The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014648.3 | NP_055463.1 | 29 - 30 | 3690 | 144 |
XM_005247914.3 | XP_005247971.1 | 29 - 30 | 3489 | 144 | |
XM_005247915.3 | XP_005247972.1 | 29 - 30 | 3459 | 144 | |
XM_005247916.3 | XP_005247973.1 | 30 - 31 | 3631 | 144 | |
XM_005247917.3 | XP_005247974.1 | 29 - 30 | 3420 | 144 | |
XM_005247918.3 | XP_005247975.1 | 29 - 30 | 3907 | 144 | |
XM_005247919.3 | XP_005247976.1 | 28 - 29 | 3041 | 144 | |
XM_017007540.1 | XP_016863029.1 | 29 - 30 | 3536 | 144 | |
GenBank mRNA | AB014575.1 | - | 29 - 30 | 3397 | 144 |
AF279370.1 | - | 14 - 15 | 1349 | 144 | |
AK023138.1 | - | 29 - 30 | 3458 | 144 | |
AK294967.1 | - | 18 - 19 | 1753 | 144 | |
AY227652.1 | - | 16 - 17 | 1601 | 144 | |
AY227654.1 | - | 27 - 28 | 4126 | 144 | |
AY495710.1 | - | 4 - 5 | 471 | 144 | |
BC063882.1 | - | 29 - 30 | 3443 | 144 |