The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014648.3 | NP_055463.1 | 16 - 17 | 2379 | 134 |
XM_005247914.3 | XP_005247971.1 | 16 - 17 | 2178 | 134 | |
XM_005247915.3 | XP_005247972.1 | 16 - 17 | 2148 | 134 | |
XM_005247916.3 | XP_005247973.1 | 17 - 18 | 2320 | 134 | |
XM_005247917.3 | XP_005247974.1 | 16 - 17 | 2109 | 134 | |
XM_005247918.3 | XP_005247975.1 | 16 - 17 | 2596 | 134 | |
XM_005247919.3 | XP_005247976.1 | 15 - 16 | 1730 | 134 | |
XM_017007540.1 | XP_016863029.1 | 16 - 17 | 2225 | 134 | |
GenBank mRNA | AB014575.1 | - | 16 - 17 | 2086 | 134 |
AK023138.1 | - | 16 - 17 | 2147 | 134 | |
AK294967.1 | - | 5 - 6 | 442 | 134 | |
AK303942.1 | - | 3 - 4 | 582 | 134 | |
AY227652.1 | - | 3 - 4 | 290 | 134 | |
AY227653.1 | - | 7 - 8 | 1142 | 134 | |
BC063882.1 | - | 16 - 17 | 2132 | 134 |