The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020890.2 | NP_065941.2 | 13 - 14 | 1907 | 73 |
XM_006713716.3 | XP_006713779.1 | 13 - 14 | 1691 | 73 | |
XM_006713717.2 | XP_006713780.1 | 14 - 15 | 1803 | 73 | |
XM_011513056.1 | XP_011511358.1 | 13 - 14 | 1651 | 73 | |
XM_011513057.2 | XP_011511359.1 | 14 - 15 | 1772 | 73 | |
GenBank mRNA | AB040957.1 | - | 13 - 14 | 1634 | 73 |
AF334474.1 | - | 13 - 14 | 1706 | 73 | |
AK022912.1 | - | 4 - 5 | 518 | 73 | |
AK056933.1 | - | 13 - 14 | 1716 | 73 | |
AK308315.1 | - | 13 - 14 | 1653 | 73 | |
AL713689.1 | - | 13 - 14 | 1702 | 73 | |
BC063433.1 | - | 13 - 14 | 1698 | 73 | |
BC082961.1 | - | 13 - 14 | 1660 | 73 | |
BC130564.1 | - | 13 - 14 | 1674 | 73 | |
BC136371.1 | - | 13 - 14 | 1674 | 73 |