Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARMCX2
Entrez Gene ID
9823
Gene Name
armadillo repeat containing, X-linked 2
Gene Aliases
ALEX2, GASP9
Chromosome Location
Chr.X: 101655281 - 101659891 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014782.6 NP_055597.1 3 - 4 304 95
XM_005278110.1 XP_005278167.1 4 - 5 386 95
XM_005278114.1 XP_005278171.1 4 - 5 374 95
XM_011531071.1 XP_011529373.1 4 - 5 430 95
XM_011531072.1 XP_011529374.1 4 - 5 397 95
XM_017029990.1 XP_016885479.1 4 - 5 385 95
GenBank mRNA BC012541.1 - 3 - 4 260 95

View Full Product Details