Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIRT5
Entrez Gene ID
23408
Gene Name
sirtuin 5
Gene Aliases
SIR2L5
Chromosome Location
Chr.6: 13574222 - 13615158 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193267.2 NP_001180196.1 2 - 3 255 79
NM_001242827.1 NP_001229756.1 2 - 3 255 79
NM_012241.4 NP_036373.1 2 - 3 277 79
NM_031244.3 NP_112534.1 2 - 3 255 79
XM_005248968.4 XP_005249025.1 2 - 3 192 79
XM_011514444.2 XP_011512746.1 2 - 3 185 79
XM_017010620.1 XP_016866109.1 1 - 2 155 79
XM_017010621.1 XP_016866110.1 2 - 3 192 79
XM_017010622.1 XP_016866111.1 2 - 3 244 79
XM_017010623.1 XP_016866112.1 2 - 3 374 79
XM_017010624.1 XP_016866113.1 1 - 2 155 79
XM_017010625.1 XP_016866114.1 1 - 2 155 79
GenBank mRNA AF083110.1 - 2 - 3 238 79
AJ420496.1 - 2 - 3 184 79
AK000355.1 - 2 - 3 183 79
AK294162.1 - 2 - 3 183 79
AK302467.1 - 2 - 3 190 79
BC000126.1 - 2 - 3 261 79
BX538030.1 - 2 - 3 202 79
DC314190.1 - 2 - 3 277 79

View Full Product Details