The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003786.3 | NP_003777.2 | 25 - 26 | 3790 | 93 |
XM_005257763.3 | XP_005257820.1 | 24 - 25 | 3612 | 93 | |
XM_011525422.2 | XP_011523724.1 | 24 - 25 | 3717 | 93 | |
XM_011525423.1 | XP_011523725.1 | 25 - 26 | 3909 | 93 | |
XM_011525424.2 | XP_011523726.1 | 20 - 21 | 3140 | 93 | |
XM_011525425.2 | XP_011523727.1 | 24 - 25 | 3617 | 93 | |
XM_017025265.1 | XP_016880754.1 | 21 - 22 | 3653 | 93 | |
GenBank mRNA | AB010887.1 | - | 25 - 26 | 3724 | 93 |
AB208954.1 | - | 25 - 26 | 3730 | 93 | |
AF009670.1 | - | 25 - 26 | 3783 | 93 | |
AF083552.1 | - | 25 - 26 | 3727 | 93 | |
AF085690.1 | - | 25 - 26 | 3746 | 93 | |
AF085691.1 | - | 24 - 25 | 3895 | 93 | |
AF085692.1 | - | 24 - 25 | 3950 | 93 | |
AF104943.1 | - | 25 - 26 | 3714 | 93 | |
BC137347.1 | - | 25 - 26 | 3741 | 93 | |
BC137348.1 | - | 25 - 26 | 3741 | 93 | |
Y17151.2 | - | 25 - 26 | 3752 | 93 |