The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286229.1 | NP_001273158.1 | 8 - 9 | 1031 | 149 |
NM_001286230.1 | NP_001273159.1 | 9 - 10 | 1135 | 149 | |
NM_016448.3 | NP_057532.3 | 10 - 11 | 1261 | 149 | |
XM_011509614.1 | XP_011507916.1 | 10 - 11 | 965 | 149 | |
GenBank mRNA | AF195765.1 | - | 10 - 11 | 1025 | 149 |
AF345896.1 | - | 10 - 11 | 1046 | 149 | |
AK000742.1 | - | 10 - 11 | 1079 | 149 | |
AK027651.1 | - | 8 - 9 | 738 | 149 | |
AK292343.1 | - | 10 - 11 | 1133 | 149 | |
AK303346.1 | - | 9 - 10 | 938 | 149 | |
BC033297.1 | - | 10 - 11 | 1002 | 149 | |
BC033540.1 | - | 10 - 11 | 1043 | 149 | |
DQ641253.1 | - | 10 - 11 | 923 | 149 |