The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164821.1 | NP_001158293.1 | 9 - 10 | 1308 | 111 |
GenBank mRNA | AK307877.1 | - | 9 - 10 | 1308 | 111 |