Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GAS2L1
Entrez Gene ID
10634
Gene Name
growth arrest specific 2 like 1
Gene Aliases
GAR22
Chromosome Location
Chr.22: 29306618 - 29312789 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006478.4 NP_006469.2 1 - 2 159 138

View Full Product Details