The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242318.2 | NP_001229247.1 | 12 - 13 | 1468 | 86 |
NM_002603.3 | NP_002594.1 | 12 - 13 | 1317 | 86 | |
XM_011517540.2 | XP_011515842.1 | 13 - 14 | 1262 | 86 | |
XM_017013538.1 | XP_016869027.1 | 12 - 13 | 1829 | 86 | |
GenBank mRNA | AK290801.1 | - | 12 - 13 | 1276 | 86 |
AK292680.1 | - | 12 - 13 | 1265 | 86 | |
BC126360.1 | - | 12 - 13 | 1309 | 86 | |
U67932.1 | - | 12 - 13 | 1320 | 86 |