Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDE7A
Entrez Gene ID
5150
Gene Name
phosphodiesterase 7A
Gene Aliases
HCP1, PDE7
Chromosome Location
Chr.8: 65714334 - 65841734 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242318.2 NP_001229247.1 7 - 8 929 69
NM_002603.3 NP_002594.1 7 - 8 778 69
XM_011517540.2 XP_011515842.1 8 - 9 723 69
XM_017013538.1 XP_016869027.1 7 - 8 1290 69
GenBank mRNA AF332652.1 - 7 - 8 703 69
AK290801.1 - 7 - 8 737 69
AK292680.1 - 7 - 8 726 69
BC126360.1 - 7 - 8 770 69
U67932.1 - 7 - 8 781 69

View Full Product Details