The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033046.3 | NP_001028218.1 | 5 - 6 | 471 | 80 |
NM_001100407.2 | NP_001093877.1 | 6 - 7 | 521 | 80 | |
NM_001100408.2 | NP_001093878.1 | 4 - 5 | 429 | 80 | |
NM_001193653.1 | NP_001180582.1 | 6 - 7 | 741 | 80 | |
NM_001193654.1 | NP_001180583.1 | 5 - 6 | 691 | 80 | |
NM_001193655.1 | NP_001180584.1 | 5 - 6 | 635 | 80 | |
NM_001193657.1 | NP_001180586.1 | 6 - 7 | 688 | 80 | |
NR_036514.1 | 4 - 5 | 397 | 80 | ||
NR_036516.1 | 4 - 5 | 348 | 80 | ||
NR_036517.1 | 3 - 4 | 306 | 80 | ||
NR_036518.1 | 2 - 3 | 232 | 80 | ||
XM_006722293.2 | XP_006722356.1 | 4 - 5 | 1196 | 80 | |
XM_011523606.2 | XP_011521908.1 | 4 - 5 | 1376 | 80 | |
XM_017025072.1 | XP_016880561.1 | 4 - 5 | 1376 | 80 | |
XM_017025073.1 | XP_016880562.1 | 3 - 4 | 1334 | 80 | |
XM_017025074.1 | XP_016880563.1 | 5 - 6 | 469 | 80 | |
XM_017025075.1 | XP_016880564.1 | 5 - 6 | 1238 | 80 | |
XM_017025076.1 | XP_016880565.1 | 6 - 7 | 466 | 80 | |
XM_017025077.1 | XP_016880566.1 | 5 - 6 | 630 | 80 | |
XM_017025078.1 | XP_016880567.1 | 4 - 5 | 427 | 80 | |
GenBank mRNA | AK027743.1 | - | 2 - 3 | 172 | 80 |
AK054876.1 | - | 4 - 5 | 427 | 80 | |
AK056342.1 | - | 4 - 5 | 304 | 80 | |
AK074950.1 | - | 6 - 7 | 618 | 80 | |
AK090484.1 | - | 4 - 5 | 2542 | 80 | |
AK225389.1 | - | 5 - 6 | 377 | 80 | |
AU280357.1 | - | 5 - 6 | 416 | 80 | |
BC003595.1 | - | 5 - 6 | 355 | 80 | |
BC004171.2 | - | 5 - 6 | 355 | 80 | |
BG716477.1 | - | 5 - 6 | 571 | 80 | |
BM147030.1 | - | 4 - 5 | 253 | 80 | |
BM801484.1 | - | 6 - 7 | 454 | 80 | |
CN260096.1 | - | 4 - 5 | 255 | 80 | |
DA008508.1 | - | 3 - 4 | 203 | 80 | |
DA342707.1 | - | 4 - 5 | 419 | 80 | |
DA500882.1 | - | 4 - 5 | 353 | 80 | |
DA663677.1 | - | 6 - 7 | 443 | 80 | |
DA716299.1 | - | 5 - 6 | 475 | 80 | |
DB113745.1 | - | 4 - 5 | 252 | 80 |