The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018346.2 | NP_060816.1 | 5 - 6 | 993 | 113 |
GenBank mRNA | AK002026.1 | - | 5 - 6 | 964 | 113 |
AK022105.1 | - | 5 - 6 | 933 | 113 | |
AK222472.1 | - | 5 - 6 | 944 | 113 | |
BC005854.2 | - | 4 - 5 | 745 | 113 | |
BC050538.1 | - | 5 - 6 | 924 | 113 |