The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319945.1 | NP_001306874.1 | 2 - 3 | 562 | 67 |
NM_138387.3 | NP_612396.1 | 2 - 3 | 562 | 67 | |
NR_028581.1 | 3 - 4 | 756 | 67 | ||
NR_028582.1 | 2 - 3 | 621 | 67 | ||
XM_011525473.2 | XP_011523775.1 | 3 - 4 | 316 | 67 | |
XM_011525474.2 | XP_011523776.1 | 3 - 4 | 785 | 67 | |
XM_017025335.1 | XP_016880824.1 | 2 - 3 | 307 | 67 | |
GenBank mRNA | BC002494.2 | - | 2 - 3 | 360 | 67 |
BC021574.1 | - | 2 - 3 | 542 | 67 |