The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319945.1 | NP_001306874.1 | 4 - 5 | 795 | 64 |
NM_138387.3 | NP_612396.1 | 5 - 6 | 914 | 64 | |
NR_028581.1 | 6 - 7 | 1108 | 64 | ||
NR_028582.1 | 5 - 6 | 973 | 64 | ||
XM_011525473.2 | XP_011523775.1 | 6 - 7 | 668 | 64 | |
XM_011525474.2 | XP_011523776.1 | 6 - 7 | 1137 | 64 | |
XM_017025335.1 | XP_016880824.1 | 5 - 6 | 659 | 64 | |
GenBank mRNA | BC002494.2 | - | 5 - 6 | 712 | 64 |
BC021574.1 | - | 5 - 6 | 894 | 64 |