The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142610.1 | NP_001136082.1 | 11 - 12 | 1346 | 142 |
NM_014683.3 | NP_055498.3 | 11 - 12 | 1346 | 142 | |
XM_011524087.2 | XP_011522389.1 | 7 - 8 | 643 | 142 | |
XM_017025424.1 | XP_016880913.1 | 11 - 12 | 1234 | 142 | |
XM_017025425.1 | XP_016880914.1 | 11 - 12 | 1234 | 142 | |
XM_017025426.1 | XP_016880915.1 | 11 - 12 | 1234 | 142 | |
XM_017025427.1 | XP_016880916.1 | 9 - 10 | 688 | 142 | |
XM_017025428.1 | XP_016880917.1 | 9 - 10 | 688 | 142 | |
GenBank mRNA | AB014523.1 | - | 11 - 12 | 1337 | 142 |
AK098316.1 | - | 10 - 11 | 971 | 142 | |
AK314345.1 | - | 11 - 12 | 869 | 142 | |
BC034988.1 | - | 12 - 13 | 911 | 142 |