The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142610.1 | NP_001136082.1 | 19 - 20 | 2505 | 127 |
NM_014683.3 | NP_055498.3 | 19 - 20 | 2505 | 127 | |
XM_011524087.2 | XP_011522389.1 | 15 - 16 | 1802 | 127 | |
XM_017025424.1 | XP_016880913.1 | 19 - 20 | 2393 | 127 | |
XM_017025425.1 | XP_016880914.1 | 19 - 20 | 2393 | 127 | |
XM_017025426.1 | XP_016880915.1 | 19 - 20 | 2393 | 127 | |
XM_017025427.1 | XP_016880916.1 | 17 - 18 | 1847 | 127 | |
XM_017025428.1 | XP_016880917.1 | 17 - 18 | 1847 | 127 | |
GenBank mRNA | AB014523.1 | - | 19 - 20 | 2496 | 127 |
AK098316.1 | - | 18 - 19 | 2130 | 127 | |
AK314345.1 | - | 19 - 20 | 2028 | 127 | |
BC034988.1 | - | 20 - 21 | 2070 | 127 |