Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCL9
Entrez Gene ID
607
Gene Name
B-cell CLL/lymphoma 9
Gene Aliases
LGS
Chromosome Location
Chr.1: 147541394 - 147626219 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004326.3 NP_004317.2 8 - 9 3549 63
XM_005272971.4 XP_005273028.1 8 - 9 3554 63
XM_005277417.3 XP_005277474.1 8 - 9 3578 63
XM_005277418.3 XP_005277475.1 8 - 9 3434 63
XM_017002026.1 XP_016857515.1 8 - 9 3332 63
GenBank mRNA BC116451.1 - 5 - 6 2944 63
Y13620.1 - 8 - 9 3637 63

View Full Product Details