Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCL9
Entrez Gene ID
607
Gene Name
B-cell CLL/lymphoma 9
Gene Aliases
LGS
Chromosome Location
Chr.1: 147541394 - 147626219 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004326.3 NP_004317.2 9 - 10 3811 73
XM_005272971.4 XP_005273028.1 9 - 10 3816 73
XM_005277418.3 XP_005277475.1 9 - 10 3696 73
XM_017002026.1 XP_016857515.1 9 - 10 3594 73
GenBank mRNA BC116451.1 - 6 - 7 3206 73
Y13620.1 - 9 - 10 3899 73

View Full Product Details