The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052898.1 | NP_443130.1 | 3 - 3 | 3208 | 91 |
GenBank mRNA | AB067476.1 | - | 3 - 3 | 2742 | 91 |
AY534241.1 | - | 3 - 3 | 3208 | 91 |