Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- MLXIPL
- Entrez Gene ID
-
51085
- Gene Name
- MLX interacting protein like
- Gene Aliases
-
CHREBP, MIO, MONDOB, WBSCR14, WS-bHLH, bHLHd14
- Chromosome Location
-
Chr.7: 73593194 - 73624543 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC012925.1 |
- |
4 - 5 |
550 |
132 |
View Full Product Details