The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145838.1 | NP_001139310.1 | 3 - 4 | 501 | 75 |
NM_001145839.1 | NP_001139311.1 | 5 - 6 | 732 | 75 | |
NM_014637.3 | NP_055452.3 | 5 - 6 | 732 | 75 | |
XM_006716484.2 | XP_006716547.2 | 5 - 6 | 704 | 75 | |
XM_011517626.1 | XP_011515928.1 | 5 - 6 | 686 | 75 | |
XM_011517627.2 | XP_011515929.1 | 5 - 6 | 622 | 75 | |
XM_011517628.2 | XP_011515930.1 | 4 - 5 | 839 | 75 | |
GenBank mRNA | AK304716.1 | - | 5 - 6 | 646 | 75 |
BC036116.1 | - | 5 - 6 | 646 | 75 | |
BC043498.2 | - | 5 - 6 | 631 | 75 | |
BX537854.1 | - | 3 - 4 | 393 | 75 | |
BX647507.1 | - | 5 - 6 | 651 | 75 | |
CR456910.1 | - | 3 - 4 | 463 | 75 | |
D13634.1 | - | 4 - 5 | 480 | 75 |