Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MTFR1
Entrez Gene ID
9650
Gene Name
mitochondrial fission regulator 1
Gene Aliases
CHPPR, FAM54A2
Chromosome Location
Chr.8: 65643873 - 65771252 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145838.1 NP_001139310.1 4 - 5 744 142
NM_001145839.1 NP_001139311.1 6 - 7 975 142
NM_014637.3 NP_055452.3 6 - 7 975 142
XM_006716484.2 XP_006716547.2 6 - 7 947 142
XM_011517626.1 XP_011515928.1 6 - 7 929 142
XM_011517627.2 XP_011515929.1 6 - 7 865 142
XM_011517628.2 XP_011515930.1 5 - 6 1082 142
GenBank mRNA AK298945.1 - 5 - 6 473 142
AK304716.1 - 6 - 7 889 142
AK316202.1 - 5 - 6 515 142
BC036116.1 - 6 - 7 889 142
BC043498.2 - 6 - 7 874 142
BX537854.1 - 4 - 5 636 142
BX647507.1 - 6 - 7 894 142
CR456910.1 - 4 - 5 706 142
D13634.1 - 5 - 6 723 142

View Full Product Details