The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105539.2 | NP_001099009.1 | 1 - 2 | 1567 | 64 |
NM_023929.4 | NP_076418.3 | 1 - 2 | 1567 | 64 | |
GenBank mRNA | BC127097.1 | - | 2 - 3 | 849 | 64 |
BC127098.1 | - | 2 - 3 | 579 | 64 | |
BC148254.1 | - | 2 - 3 | 537 | 64 |