Product Details

Assay Reference Genome
Location

Chr.19:18785515 on build GRCh38
Cytoband
19p13.11
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
1311
Gene Symbol
COMP
Gene Name
cartilage oligomeric matrix protein
Gene Aliases
EDM1, EPD1, MED, PSACH, THBS5, TSP5
Location
Chr.19:18782773-18791305 on build GRCh38
Assay Gene Location
Overlaps Intron 14 - Intron 15
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
COMP NM_000095.2 NP_000086.2
AB086984.1 BAC53888.1
AK074508.1 BAC11031.1
AK223216.1 BAD96936.1
AK290595.1
AK296586.1
AK297798.1
AK297814.1
BC110847.1 AAI10848.1
BC125092.1
L32137.1 AAA57253.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1151217 Chr19:18627369 - 19284914 on Build GRCh38 Deletion KLHL26 NR2C2AP SUGP1 TM6SF2 BORCS8-MEF2B DDX49 CRTC1 RFXANK SLC25A42 SUGP2 COPE HOMER3 GDF1 COMP LOC102724360 MEF2B HAPLN4 BORCS8 TMEM161A NCAN ARMC6 CERS1 UPF1
nsv952422 Chr19:18606691 - 18807891 on Build GRCh38 Deletion KLHL26 COMP TMEM59L CRTC1 CRLF1
nsv833777 Chr19:18765126 - 18920267 on Build GRCh38 Loss GDF1 COMP DDX49 CRTC1 CERS1 COPE UPF1

View Full Product Details