The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303272.1 | NP_001290201.1 | 9 - 10 | 1201 | 68 |
NM_015462.4 | NP_056277.2 | 10 - 11 | 1259 | 68 | |
GenBank mRNA | AK023702.1 | - | 10 - 11 | 1149 | 68 |
AK299483.1 | - | 9 - 10 | 1091 | 68 | |
AY598333.1 | - | 10 - 11 | 1144 | 68 | |
BC001726.2 | - | 2 - 3 | 213 | 68 | |
BC064404.1 | - | 10 - 11 | 1150 | 68 |