Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NOL11
Entrez Gene ID
25926
Gene Name
nucleolar protein 11
Gene Aliases
-
Chromosome Location
Chr.17: 67717833 - 67744208 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015462.4 NP_056277.2 4 - 5 572 130
GenBank mRNA AK023702.1 - 4 - 5 462 130
AL110271.1 - 1 - 2 167 130
AY598333.1 - 4 - 5 457 130
BC064404.1 - 4 - 5 463 130

View Full Product Details