The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021029.5 | NP_066357.2 | 4 - 5 | 454 | 135 |
GenBank mRNA | AW150895.1 | - | 1 - 2 | 135 | |
BG504049.1 | - | 4 - 5 | 333 | 135 |