Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAD51D
Entrez Gene ID
5892
Gene Name
RAD51 paralog D
Gene Aliases
BROVCA4, R51H3, RAD51L3, TRAD
Chromosome Location
Chr.17: 35099792 - 35119869 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002878.3 NP_002869.3 3 - 4 520 91
GenBank mRNA AB013341.1 - 3 - 4 370 91
AB018360.1 - 2 - 3 167 91
AK302450.1 - 3 - 4 414 91
BC014422.1 - 3 - 4 399 91
BI916871.1 - 3 - 4 520 91
BX647297.1 - 3 - 4 467 91
Y15572.1 - 3 - 4 383 91

View Full Product Details