The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000350.2 | NP_000341.2 | 4 - 5 | 547 | 119 |
GenBank mRNA | AB210040.1 | - | 4 - 5 | 533 | 119 |
AF000148.1 | - | 4 - 5 | 530 | 119 | |
AF001945.1 | - | 4 - 5 | 524 | 119 | |
AK295818.1 | - | 4 - 5 | 523 | 119 | |
U88667.1 | - | 4 - 5 | 524 | 119 |