Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNX7
Entrez Gene ID
51375
Gene Name
sorting nexin 7
Gene Aliases
-
Chromosome Location
Chr.1: 98661344 - 98760500 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017001425.1 XP_016856914.1 1 - 2 210 89
XM_017001428.1 XP_016856917.1 1 - 2 97 89

View Full Product Details