Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DTL
Entrez Gene ID
51514
Gene Name
denticleless E3 ubiquitin protein ligase homolog
Gene Aliases
CDT2, DCAF2, L2DTL, RAMP
Chromosome Location
Chr.1: 212035553 - 212105006 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286229.1 NP_001273158.1 1 - 2 390 63
NM_001286230.1 NP_001273159.1 1 - 2 390 63
GenBank mRNA AK027651.1 - 1 - 2 97 63
AK303346.1 - 1 - 2 193 63

View Full Product Details