The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002207.2 | NP_002198.2 | 9 - 10 | 1093 | 82 |
GenBank mRNA | BC030198.1 | - | 9 - 10 | 1300 | 82 |
D25303.1 | - | 10 - 11 | 1091 | 82 | |
L24158.1 | - | 9 - 10 | 935 | 82 |