Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ITGA9
Entrez Gene ID
3680
Gene Name
integrin subunit alpha 9
Gene Aliases
ALPHA-RLC, ITGA4L, RLC
Chromosome Location
Chr.3: 37452322 - 37819790 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002207.2 NP_002198.2 11 - 12 1293 140
GenBank mRNA BC030198.1 - 11 - 12 1500 140
D25303.1 - 12 - 13 1291 140
L24158.1 - 11 - 12 1135 140

View Full Product Details