The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130964.1 | NP_001124436.1 | 13 - 14 | 2449 | 58 |
NM_006225.3 | NP_006216.2 | 13 - 14 | 2156 | 58 | |
NR_024071.1 | 12 - 13 | 2526 | 58 | ||
XM_017006622.1 | XP_016862111.1 | 13 - 14 | 2419 | 58 | |
GenBank mRNA | AB209862.1 | - | 13 - 14 | 2093 | 58 |
AK090774.1 | - | 13 - 14 | 2238 | 58 | |
AK098690.1 | - | 10 - 11 | 1439 | 58 | |
AK292324.1 | - | 13 - 14 | 2156 | 58 | |
BC050382.1 | - | 13 - 14 | 2120 | 58 | |
BX647927.1 | - | 12 - 13 | 2260 | 58 | |
U09117.1 | - | 13 - 14 | 2126 | 58 |