The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001433.3 | NP_001424.3 | 6 - 7 | 587 | 62 |
XM_017024347.1 | XP_016879836.1 | 6 - 7 | 1941 | 62 | |
GenBank mRNA | AB209869.1 | - | 6 - 7 | 570 | 62 |
AF059198.1 | - | 6 - 7 | 570 | 62 | |
AK292403.1 | - | 6 - 7 | 613 | 62 | |
BC130405.1 | - | 6 - 7 | 520 | 62 | |
BC130407.1 | - | 6 - 7 | 520 | 62 |