The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001316990.1 | NP_001303919.1 | 15 - 16 | 2170 | 85 |
NM_138363.2 | NP_612372.1 | 16 - 17 | 2276 | 85 | |
NR_133644.1 | 16 - 17 | 2299 | 85 | ||
XM_005257779.3 | XP_005257836.1 | 16 - 17 | 2252 | 85 | |
XM_005257780.2 | XP_005257837.1 | 15 - 16 | 1895 | 85 | |
XM_006722166.3 | XP_006722229.1 | 16 - 17 | 2034 | 85 | |
XM_017025287.1 | XP_016880776.1 | 16 - 17 | 3255 | 85 | |
XM_017025288.1 | XP_016880777.1 | 15 - 16 | 1928 | 85 | |
GenBank mRNA | AK290587.1 | - | 16 - 17 | 2016 | 85 |
AK297409.1 | - | 15 - 16 | 1871 | 85 | |
AL832822.1 | - | 16 - 17 | 1952 | 85 | |
BC009518.1 | - | 16 - 17 | 1972 | 85 |