The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042663.1 | NP_001036128.1 | 12 - 13 | 1407 | 73 |
NM_001042664.1 | NP_001036129.1 | 11 - 12 | 1361 | 73 | |
NM_001042665.1 | NP_001036130.1 | 11 - 12 | 1344 | 73 | |
NM_001265592.1 | NP_001252521.1 | 12 - 13 | 1440 | 73 | |
NM_001265593.1 | NP_001252522.1 | 11 - 12 | 1371 | 73 | |
NM_001265594.1 | NP_001252523.1 | 11 - 12 | 1375 | 73 | |
NM_020631.4 | NP_065682.2 | 11 - 12 | 1290 | 73 | |
NM_198681.3 | NP_941374.2 | 12 - 13 | 1919 | 73 | |
GenBank mRNA | AB018263.2 | - | 12 - 13 | 1395 | 73 |
AB097001.1 | - | 10 - 11 | 1136 | 73 | |
AK024676.1 | - | 11 - 12 | 1289 | 73 | |
AK096347.1 | - | 12 - 13 | 1867 | 73 | |
AK125550.1 | - | 12 - 13 | 1373 | 73 | |
AK131074.1 | - | 9 - 10 | 1614 | 73 | |
AK294875.1 | - | 12 - 13 | 1440 | 73 | |
AK299523.1 | - | 11 - 12 | 1371 | 73 | |
AK301239.1 | - | 5 - 6 | 370 | 73 | |
BC015231.1 | - | 11 - 12 | 1351 | 73 | |
BC036671.2 | - | 12 - 13 | 1920 | 73 |