The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042663.1 | NP_001036128.1 | 6 - 7 | 568 | 72 |
NM_001042664.1 | NP_001036129.1 | 5 - 6 | 522 | 72 | |
NM_001042665.1 | NP_001036130.1 | 5 - 6 | 505 | 72 | |
NM_001265592.1 | NP_001252521.1 | 6 - 7 | 601 | 72 | |
NM_001265593.1 | NP_001252522.1 | 5 - 6 | 532 | 72 | |
NM_001265594.1 | NP_001252523.1 | 5 - 6 | 536 | 72 | |
NM_020631.4 | NP_065682.2 | 5 - 6 | 451 | 72 | |
NM_198681.3 | NP_941374.2 | 6 - 7 | 1080 | 72 | |
GenBank mRNA | AB018263.2 | - | 6 - 7 | 556 | 72 |
AB097001.1 | - | 4 - 5 | 297 | 72 | |
AK024676.1 | - | 5 - 6 | 450 | 72 | |
AK096347.1 | - | 6 - 7 | 1028 | 72 | |
AK125550.1 | - | 6 - 7 | 534 | 72 | |
AK299523.1 | - | 5 - 6 | 532 | 72 | |
BC015231.1 | - | 5 - 6 | 512 | 72 | |
BC036671.2 | - | 6 - 7 | 1081 | 72 | |
DA091930.1 | - | 5 - 6 | 536 | 72 | |
DB093234.1 | - | 5 - 6 | 451 | 72 |