Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TANC1
Entrez Gene ID
85461
Gene Name
tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 1
Gene Aliases
ROLSB, TANC
Chromosome Location
Chr.2: 158968600 - 159232659 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145909.1 NP_001139381.1 14 - 15 2763 96
NM_033394.2 NP_203752.2 14 - 15 2787 96
XM_005246918.4 XP_005246975.1 13 - 14 2551 96
XM_006712810.3 XP_006712873.1 13 - 14 2674 96
XM_006712812.3 XP_006712875.1 13 - 14 2675 96
XM_006712814.3 XP_006712877.1 10 - 11 1970 96
XM_011512048.2 XP_011510350.1 16 - 17 2965 96
XM_011512051.2 XP_011510353.1 15 - 16 2830 96
XM_011512053.2 XP_011510355.1 14 - 15 2720 96
XM_011512055.2 XP_011510357.1 14 - 15 2756 96
XM_011512058.2 XP_011510360.1 12 - 13 2462 96
XM_011512059.2 XP_011510361.1 10 - 11 2323 96
XM_011512062.2 XP_011510364.1 8 - 9 2000 96
XM_017005141.1 XP_016860630.1 13 - 14 2579 96
XM_017005142.1 XP_016860631.1 15 - 16 3184 96
XM_017005143.1 XP_016860632.1 13 - 14 2869 96
XM_017005144.1 XP_016860633.1 13 - 14 2558 96
XM_017005145.1 XP_016860634.1 16 - 17 2999 96
XM_017005146.1 XP_016860635.1 15 - 16 2810 96
XM_017005147.1 XP_016860636.1 14 - 15 2759 96
XM_017005148.1 XP_016860637.1 15 - 16 2925 96
XM_017005149.1 XP_016860638.1 12 - 13 2477 96
XM_017005150.1 XP_016860639.1 12 - 13 2459 96
XM_017005151.1 XP_016860640.1 12 - 13 2318 96
XM_017005152.1 XP_016860641.1 12 - 13 2315 96
XM_017005153.1 XP_016860642.1 11 - 12 2264 96
XM_017005154.1 XP_016860643.1 14 - 15 2681 96
XM_017005155.1 XP_016860644.1 12 - 13 2411 96
XM_017005156.1 XP_016860645.1 10 - 11 2000 96
XM_017005157.1 XP_016860646.1 11 - 12 2177 96
XM_017005158.1 XP_016860647.1 16 - 17 2928 96
GenBank mRNA AB051515.1 - 8 - 9 1864 96
AK308899.1 - 9 - 10 1930 96
BC150597.1 - 13 - 14 2604 96
BX648327.1 - 9 - 10 2047 96

View Full Product Details