Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TANC1
- Entrez Gene ID
-
85461
- Gene Name
- tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 1
- Gene Aliases
-
ROLSB, TANC
- Chromosome Location
-
Chr.2: 158968600 - 159232659 on Build GRCh38
View Full Product Details