Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNF10
Entrez Gene ID
7556
Gene Name
zinc finger protein 10
Gene Aliases
KOX1
Chromosome Location
Chr.12: 133130628 - 133159465 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015394.4 NP_056209.2 1 - 2 127 73
GenBank mRNA BC024182.2 - 1 - 2 128 73

View Full Product Details