The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_024279.1 | - | 131 | ||
GenBank mRNA | AK094772.1 | - | - | 131 | |
AK124018.1 | - | - | 131 |